Learn More
Invitrogen™ C. botulinum BoNT-C1 Heavy Chain Monoclonal Antibody (580223)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | C. botulinum BoNT-C1 Heavy Chain |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 580223 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene Alias | BONTC1 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant C. botulinum BoNT-C1 Heavy Chain Asn866-Glu1291. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.