Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C. botulinum BoNT-C1 Heavy Chain Receptor Binding Domain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5425
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5425 100 μg
AF5425SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF5425 Supplier R&D Systems Supplier No. AF5425
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

BoNT-C1 Heavy Chain Polyclonal specifically detects BoNT-C1 Heavy Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen BoNT-C1 Heavy Chain
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. P18640
Gene Alias BoNTC1 Heavy Chain, DcD
Gene Symbols CbC4_4096
Host Species Sheep
Immunogen E. coli-derived recombinant BoNT-C1 Heavy Chain Asn866-Glu1291 Accession # P18640
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10523893
Test Specificity Detects C. botulinum BoNT-C1 Heavy Chainin direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant (r) BoNT-D-HCR is observed and 5% cross-reactivity with rBoNT-A-HCR, rBoNT-A-LC, and rBoNT-B-LC is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species C. botulinum
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.