Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C. botulinum BoNT-D Heavy Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6639
This item is not returnable.
View return policy
Description
BoNT-D Heavy Chain Monoclonal specifically detects BoNT-D Heavy Chain in C. botulinum samples. It is validated for Western Blot.Specifications
BoNT-D Heavy Chain | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P19321 | |
E. coli-derived recombinant Clostridium botulinum Neurotoxin Type D Heavy Chain Asn862-Glu1276 Accession # P19321 | |
100 μg | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
682810 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects recombinant C. botulinum BoNT-D Heavy Chain in direct ELISAs and Western blots. 100% and approximately 10% cross-reactivity with recombinant C. botulinum BoNT-C1 Heavy Chain is observed in Western blots and direct ELISAs, respectively. In direct ELISAs, no cross-reactivity with the Heavy Chains of BoNT-A, -E, or -G or the Light Chains of BoNT-A, -B, -C1, -D, -E, -F, -G is observed. | |
C. botulinum | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction