Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C. botulinum BoNT-D Light Chain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6037
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6037 100 μg
MAB6037SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6037 Supplier R&D Systems Supplier No. MAB6037
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BoNT-D Light Chain Monoclonal specifically detects BoNT-D Light Chain in C. botulinum samples. It is validated for Western Blot.

Specifications

Antigen BoNT-D Light Chain
Applications Western Blot
Classification Monoclonal
Clone 695004
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P19321
Host Species Mouse
Immunogen E. coli-derived recombinant C. botulinum BoNT-D Light Chain Thr2-Ser428 Accession # P19321
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects C. botulinum BoNT-D Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant Light Chains of BoNT-A, -B, -C1, -E, -F, -G or recombinant Heavy Chains of BoNT-A, -C1, or -D is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species C. botulinum
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.