Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C. botulinum BoNT-E Light Chain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5424
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5424 100 μg
AF5424SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF5424 Supplier R&D Systems Supplier No. AF5424
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

BoNT-E Light Chain Polyclonal specifically detects BoNT-E Light Chain in C. botulinum samples. It is validated for Western Blot.

Specifications

Antigen BoNT-E Light Chain
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P30995
Host Species Goat
Immunogen E. coli-derived recombinant C. botulinum BoNT-E Light Chain Pro2-Ile403 Accession # P30995
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects C. botulinum BoNT-E Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with rBoNT-B LC is observed and less than 1% cross-reactivity with rBoNT-C HCR and rBoNT-D HCR is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species C. botulinum
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.