Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ c-Cbl Monoclonal Antibody (S.551.7)
GREENER_CHOICE

Catalog No. PIMA514955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514955 Supplier Invitrogen™ Supplier No. MA514955
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Cbl-b or Cbl-c proteins.

Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Cbl
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S.551.7
Concentration 18.6 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene Cbl
Gene Accession No. P22681
Gene Alias 4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL
Gene Symbols Cbl
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence of human c-Cbl.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 867
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.