Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Scientific C-Clamp
SDPSCI_ED

Catalog No. S00388
Change view
Click to view available options
Holds:
Objects up to 3 in. dia.
Objects up to 4 in. dia.
Catalog No. Holds
S00388 Objects up to 4 in. dia.
S00386 Objects up to 3 in. dia.
2 options

Catalog No. S00388

Supplier: American Scientific 355403

This C-Clamp is a versatile tool used for securing and holding objects firmly in place during various tasks. It provides a strong and adjustable grip, making it essential for tasks requiring stability and precision.

  • Use this C-Clamp for clamping 2 apparatuses together or securing an apparatus to a tabletop
  • Useful for setting up rod mounted pulleys
  • The clamp is a useful tool for a variety of applications, including in physics labs and classrooms, woodworking, welding, and more
  • Cast-iron enameled frame
  • Plated screw and vise-type nut

Specifications

Autoclavable Not Autoclavable
Style C-Clamp
Quantity 1 Ea.
Type C-Clamp
Material Cast Iron
Holds Objects up to 4 in. dia.
Shape C
Color Red
Jaw Type Swivel
Sterility Non-sterile
Coating Cast Iron
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.