Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586117 Supplier Invitrogen™ Supplier No. PA586117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP beta
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Cebpb
Gene Accession No. P17676, P21272, P28033
Gene Alias AGP/EBP; C EBP; c/EBP beta; C/EBPb; C/EBPbeta; C/EBP-beta; C/EBP-related protein 2; CCAAT enhancer binding protein beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer binding protein beta; CCAAT/enhancer-binding protein beta; CEBPB; Crp2; Il6dbp; IL-6DBP; interleukin 6-dependent DNA-binding protein; interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nfil6; Nf-il6; NF-M; nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; PP9092; Sfb; SF-B; Silencer factor B; TCF5; TCF-5; transcription factor 5; transcription factor C/EBP beta
Gene Symbols Cebpb
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 200-250 of Human C/EBP-beta.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1051, 12608, 24253
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.