Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C/EBP beta Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120052
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, HepG2, Mouse skeletal muscle, Rat liver, Rat skeletal muscle. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: AELKAEPGFE PADCKRKEEA GAPGGGAGMA AGFPYALRAY LGYQAV.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.
Specifications
C/EBP beta | |
Polyclonal | |
Unconjugated | |
Cebpb | |
AGP/EBP; C EBP; c/EBP beta; C/EBPb; C/EBPbeta; C/EBP-beta; C/EBP-related protein 2; CCAAT enhancer binding protein beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer binding protein beta; CCAAT/enhancer-binding protein beta; CEBPB; Crp2; Il6dbp; IL-6DBP; interleukin 6-dependent DNA-binding protein; interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nfil6; Nf-il6; NF-M; nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; PP9092; Sfb; SF-B; Silencer factor B; TCF5; TCF-5; transcription factor 5; transcription factor C/EBP beta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1051, 12608, 24253 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.84 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17676, P21272, P28033 | |
Cebpb | |
Recombinant fusion protein containing a sequence corresponding to amino acids 171-216 of human C/EBPB (NP_0051852). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction