Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C/EBP beta Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563351
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3351 Supplier Bethyl Laboratories, Inc Supplier No. A302739A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Important transcription factor regulating the expression of genes involved in immune and inflammatory responses (PubMed:9374525, PubMed:12048245, PubMed:18647749). Plays also a significant role in adipogenesis, as well as in the gluconeogenic pathway, liver regeneration, and hematopoiesis. The consensus recognition site is 5′-T[TG]NNGNAA[TG]-3′.

Specifications

Antigen C/EBP beta
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CEBPB
Gene Accession No. P17676
Gene Alias C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAP, LIP, Liver activator protein, Liver-enriched inhibitory protein, liver-enriched transcriptional activator protein, NFIL6, NF-IL6, nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5, TCF-5, transcription factor 5, transcription factor C/EBP beta
Gene Symbols CEBPB
Host Species Rabbit
Immunogen Between 293 and 343
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1051
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.