Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP delta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534466
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534466 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534466 Supplier Invitrogen™ Supplier No. PA534466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat spleen tissue lysate. PA5-34466 can be used with blocking peptide PEP-1508.

C/EBP delta are CCAAT enhancer binding proteins (CEBPs), a family of transcription factors that all contain a highly conserved, basic-leucine zipper domain at the C-terminus that is involved in dimerization and DNA binding. At least six members of the family have been isolated and characterized to date (CEBP alpha to CEBP zeta). CEBPD is a leucine zipper (LZ) DNA-binding protein that regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. CEBPD is an important transcriptional activator in the regulation of genes involved in immune and inflammatory responses and has been reported to possess many tumor suppressor-like properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP delta
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CEBPD
Gene Accession No. P49716, Q00322, Q03484
Gene Alias C EBP; c/EBP delta; C/EBPd; C/EBPdelta; C/EBP-delta; c/EBP-related protein 3; CCAAT/enhancer binding protein (C/EBP), delta; CCAAT/enhancer binding protein delta; CCAAT/enhancer-binding protein delta; CEBPD; Celf; CRP3; NF-IL6-beta; Nuclear factor NF-IL6-beta; transcription factor CELF
Gene Symbols CEBPD
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human CEBPD.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1052, 12609, 25695
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.