Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP delta/epsilon Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599597 Supplier Invitrogen™ Supplier No. PA599597
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C/EBP delta/epsilon.

C/EBP α regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. C/EBP α uses a bipartite structural motif to bind DNA. Two protein chains dimerize through a set of amphipathic α helices termed the leucine zipper. Highly basic polypeptide regions emerge from the zipper to form a linked set of DNA contact surfaces. C/EBP α appears to function exclusively in terminally-differentiated, growth-arrested cells. Additional family members include C/EBP β, C/EBP γ, C/EBP δ and C/EBP ε, all of which exhibit similar DNA-binding specificities and affinities to C/EBP α. Furthermore, C/EBP β and C/EBP δ readily form heterodimers both with each other as well as with C/EBP α.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP delta/epsilon
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEBPD
Gene Accession No. P49716, Q00322, Q15744, Q6PZD9
Gene Alias C EBP; C/EBP delta; c/EBP epsilon; C/EBPd; C/EBPdelta; C/EBP-delta; C/EBPe; C/EBPepsilon; C/EBP-epsilon; C/EBP-related protein 1; c/EBP-related protein 3; C/EPBe; CCAAT/enhancer binding protein (C/EBP), delta; CCAAT/enhancer binding protein (C/EBP), epsilon; CCAAT/enhancer binding protein , epsilon; CCAAT/enhancer binding protein delta; CCAAT/enhancer binding protein epsilon; CCAAT/enhancer-binding protein delta; CCAAT/enhancer-binding protein epsilon; CEBPD; Cebpe; Celf; CRP1; Crp3; Gm294; NF-IL6-beta; nuclear factor NF-IL6-beta; Transcription factor CELF
Gene Symbols CEBPD, CEBPE
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEBPD(Accession P49716), corresponding to amino acid residues K184-H234.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1052, 1053, 110794, 12609
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.