Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP delta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111625 Supplier Invitrogen™ Supplier No. PA5111625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKRRNQEMQQ KLVELSAENE KLHQRVEQLT RDLAGLRQFF KQLPSPPFLP AAGTADCR Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 91%.

C/EBP delta are CCAAT enhancer binding proteins (CEBPs), a family of transcription factors that all contain a highly conserved, basic-leucine zipper domain at the C-terminus that is involved in dimerization and DNA binding. At least six members of the family have been isolated and characterized to date (CEBP alpha to CEBP zeta). CEBPD is a leucine zipper (LZ) DNA-binding protein that regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. CEBPD is an important transcriptional activator in the regulation of genes involved in immune and inflammatory responses and has been reported to possess many tumor suppressor-like properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP delta
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEBPD
Gene Accession No. P49716
Gene Alias C EBP; c/EBP delta; C/EBPd; C/EBPdelta; C/EBP-delta; c/EBP-related protein 3; CCAAT/enhancer binding protein (C/EBP), delta; CCAAT/enhancer binding protein delta; CCAAT/enhancer-binding protein delta; CEBPD; Celf; CRP3; NF-IL6-beta; Nuclear factor NF-IL6-beta; transcription factor CELF
Gene Symbols CEBPD
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human C/EBP delta. Recombinant protein control fragment (Product #RP-107587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1052
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.