Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP epsilon Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582044 Supplier Invitrogen™ Supplier No. PA582044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHGTYYECEP RGGQQPLEFS GGRAGPGELG DMCEHEASID LSAYIESGEE QLLSDLFAVK PAPEARGLKG PGTPAFPHYL PPDPRPFAYP PHTFGPDRKA LGPGIYSSPG SYDPRAVAVK EEPR Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.

C/EBP alpha regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. C/EBP alpha uses a bipartite structural motif to bind DNA. Two protein chains dimerize through a set of amphipathic alpha helices termed the leucine zipper. Highly basic polypeptide regions emerge from the zipper to form a linked set of DNA contact surfaces. C/EBP alpha appears to function exclusively in terminally-differentiated, growth-arrested cells. Additional family members include C/EBP beta, C/EBP gamma, C/EBP delta and C/EBP epsilon, all of which exhibit similar DNA-binding specificities and affinities to C/EBP alpha. Furthermore, C/EBP beta and C/EBP delta readily form heterodimers both with each other as well as with C/EBP alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP epsilon
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEBPE
Gene Accession No. Q15744
Gene Alias C EBP; c/EBP epsilon; C/EBPe; C/EBPepsilon; C/EBP-epsilon; C/EBP-related protein 1; C/EPBe; CCAAT/enhancer binding protein (C/EBP), epsilon; CCAAT/enhancer binding protein , epsilon; CCAAT/enhancer binding protein epsilon; CCAAT/enhancer-binding protein epsilon; CEBPE; Crp1; Gm294
Gene Symbols CEBPE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEBPE. Recombinant protein control fragment (Product #RP-89076).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1053
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.