Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP epsilon Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121171 Supplier Invitrogen™ Supplier No. PA5121171
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: NCI-H460, Mouse spleen. The target is usually found in the following locations: Nucleus. Immunogen sequence: MSHGTYYECE PRGGQQPLEF SGGRAGPGEL GDMCEHEASI DLSAYIESGE EQLLSDLFAV KPAPEARGLK GPGTPAFPHY LPPDPRPFAY PPHTFGPDRK ALGPGIYSSP GSYDPRAVAV KEEPRGPEGS RAASRGSYNP LQYQVAHCGQ TAMHLPPTLA APGQPLRVLK APLATAAPPC SPLLKAPSPA.

C/EBP alpha regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. C/EBP alpha uses a bipartite structural motif to bind DNA. Two protein chains dimerize through a set of amphipathic alpha helices termed the leucine zipper. Highly basic polypeptide regions emerge from the zipper to form a linked set of DNA contact surfaces. C/EBP alpha appears to function exclusively in terminally-differentiated, growth-arrested cells. Additional family members include C/EBP beta, C/EBP gamma, C/EBP delta and C/EBP epsilon, all of which exhibit similar DNA-binding specificities and affinities to C/EBP alpha. Furthermore, C/EBP beta and C/EBP delta readily form heterodimers both with each other as well as with C/EBP alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP epsilon
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CEBPE
Gene Accession No. Q15744, Q6PZD9
Gene Alias C EBP; c/EBP epsilon; C/EBPe; C/EBPepsilon; C/EBP-epsilon; C/EBP-related protein 1; C/EPBe; CCAAT/enhancer binding protein (C/EBP), epsilon; CCAAT/enhancer binding protein , epsilon; CCAAT/enhancer binding protein epsilon; CCAAT/enhancer-binding protein epsilon; CEBPE; Crp1; Gm294
Gene Symbols CEBPE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human CEBPE (NP_001796.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1053, 110794
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.