Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101128 Supplier Invitrogen™ Supplier No. PA5101128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CEBPG.

The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP alpha, C/EBP beta, C/EBP gamma, and C/EBP delta, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein gamma may cooperate with Fos to bind PRE-I enhancer elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP gamma
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEBPG
Gene Accession No. P26801, P53567, P53568
Gene Alias C EBP; C/EBP; c/EBP gamma; C/EBP[g]; C/EBPgamma; CCAAT/enhancer binding protein (C/EBP), gamma; CCAAT/enhancer binding protein ,gamma; CCAAT/enhancer binding protein gamma; CCAAT/enhancer-binding protein gamma; CEBP gamma; CEBPG; CEBPRNA; G-CSF promoter element 1 binding protein; GPE1-binding protein; Gpe1bp; GPE1-BP; GPE1-BP (C/EBP gamma); Granulocyte colony-stimulating factor promoter element 1-binding protein; homolog to a human CCAAT/enhancer binding protein - gamma; Ig/EBP; IG/EBP-1; immunoglobulin enhancer-binding protein 1
Gene Symbols CEBPG
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEBPG(Accession P53567), corresponding to amino acid residues K44-V94.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1054, 12611, 25301
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.