Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP zeta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537925
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537925 Supplier Invitrogen™ Supplier No. PA537925
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

This gene belongs to the CEBP family. The encoded protein plays a role in cellular response to environmental stimuli through a transcriptional process that involves heat shock factors, conserved DNA elements (heat shock elements or HSEs) and CCAAT boxes. The protein acts as a DNA-binding transcriptional activator and regulates the heat-shock protein 70 (HSP70) promoter in a CCAAT-dependent manner. The protein is also involved in cell growth and differentiation, particularly, hematopoietic differentiation. Methylation of the promoter of this gene or mutations within the gene may be correlated with occurrence of acute myeloid leukemia (AML).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP zeta
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CEBPZ
Gene Accession No. P53569
Gene Alias AI848081; C EBP; C/EBP zeta; CBF; Cbf2; CCAAT/enhancer binding protein (C/EBP), zeta; CCAAT/enhancer binding protein alpha (C/EBP) related sequence 1; CCAAT/enhancer binding protein cebp family zeta; CCAAT/enhancer binding protein zeta; CCAAT/enhancer-binding protein zeta; CCAAT-binding factor; CCAAT-box-binding transcription factor; CEBP zeta; Cebpa-rs1; CEBPZ; core-binding factor; CTF2; HSPCBF; HSP-CBF; NOC1
Gene Symbols CEBPZ
Host Species Goat
Immunogen Peptide with sequence C-DNASFKQLKWEAERD, from the C-terminus of mouse Cebpz (aa 1005-1020).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12607
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.