Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP zeta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584099 Supplier Invitrogen™ Supplier No. PA584099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTTKTRALTV AHELLCNKPE EEKALLVQVV NKLGDPQNRI ATKASHLLET LLCKHPNMKG VVSGEVERLL FRSNISSKAQ YYAICFLNQM ALSHEESELA N Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 89%.

This gene belongs to the CEBP family. The encoded protein plays a role in cellular response to environmental stimuli through a transcriptional process that involves heat shock factors, conserved DNA elements (heat shock elements or HSEs) and CCAAT boxes. The protein acts as a DNA-binding transcriptional activator and regulates the heat-shock protein 70 (HSP70) promoter in a CCAAT-dependent manner. The protein is also involved in cell growth and differentiation, particularly, hematopoietic differentiation. Methylation of the promoter of this gene or mutations within the gene may be correlated with occurrence of acute myeloid leukemia (AML).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP zeta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEBPZ
Gene Accession No. Q03701
Gene Alias AI848081; C EBP; C/EBP zeta; CBF; Cbf2; CCAAT/enhancer binding protein (C/EBP), zeta; CCAAT/enhancer binding protein alpha (C/EBP) related sequence 1; CCAAT/enhancer binding protein cebp family zeta; CCAAT/enhancer binding protein zeta; CCAAT/enhancer-binding protein zeta; CCAAT-binding factor; CCAAT-box-binding transcription factor; CEBP zeta; Cebpa-rs1; CEBPZ; core-binding factor; CTF2; HSPCBF; HSP-CBF; NOC1
Gene Symbols CEBPZ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEBPZ. Recombinant protein control fragment (Product #RP-91656).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.