Learn More
C/EBP zeta Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303155AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
CEBPZ stimulates transcription from the HSP70 promoter.Specifications
C/EBP zeta | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q03701 | |
CEBPZ | |
Between 925 and 975 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CEBPZ | |
CBF, CBF2, CCAAT/enhancer binding protein (C/EBP), zeta, CCAAT/enhancer binding proteincebp* family zeta, CCAAT-binding factor, CCAAT-box-binding transcription factor, CTF2, HSP-CBF, NOC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10153 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.