Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C/EBP zeta Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564401
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4401 Supplier Bethyl Laboratories, Inc Supplier No. A303154A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CEBPZ stimulates transcription from the HSP70 promoter.

Specifications

Antigen C/EBP zeta
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CEBPZ
Gene Accession No. Q03701
Gene Alias CBF, CBF2, CCAAT/enhancer binding protein (C/EBP), zeta, CCAAT/enhancer binding proteincebp* family zeta, CCAAT-binding factor, CCAAT-box-binding transcription factor, CTF2, HSP-CBF, NOC1
Gene Symbols CEBPZ
Host Species Rabbit
Immunogen Between 125 and 175
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10153
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.