Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ c-ErbB2/c-Neu, Mouse, Unlabeled, Clone: 7.16.4,

Catalog No. OP16100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP16100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP16100UG Supplier MilliporeSigma™ Supplier No. OP16100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recognizes the ∽185kDa c-ErbB2/c-Neu protein in overexpressing B104-1-1 cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-ErbB2/c-Neu
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 7.16.4
Concentration Please refer to vial label for lot-specific concentration
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5, ≤0.1% Sodium Azide.
Gene Alias Anti-HER2, Anti-c-Neu, Anti-ErbB2, Anti-Erythroblastosis Virus
Host Species Mouse
Immunogen B104-1-1 cells overexpressing rat c-ErbB2/c-Neu
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 2°-8°C
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.