Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-erbB3/HER3 Polyclonal Antibody, Abbomax

Catalog No. 502204083
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4083 Supplier Abbomax Inc Supplier No. 602540
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: MCF-7 whole cell lysates Cellular location: Secreted and Cell membrane.

C-erbB3/HER3 Polyclonal antibody specifically detects C-erbB3/HER3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen C-erbB3/HER3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene ERBB3
Gene Accession No. P21860
Gene Alias avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Host Species Rabbit
Immunogen A synthetic peptide of c-erbB3 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2065
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.