Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Fos Antibody (CL6381), Novus Biologicals™
SDP

Catalog No. NB402770 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402770 25 μL
NBP276492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB402770 Supplier Novus Biologicals Supplier No. NBP27649225UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-Fos Monoclonal specifically detects c-Fos in Human samples. It is validated for Western Blot.

Specifications

Antigen c-Fos
Applications Western Blot
Classification Monoclonal
Clone CL6381
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, c-fos, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, G0/G1 switch regulatory protein 7, G0S7, proto-oncogene c-Fos, v-fos FBJ murine osteosarcoma viral oncogene homolog
Gene Symbols FOS
Host Species Mouse
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: PESEEAFTLPLLNDPEPK
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2353
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.