Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Fos Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB270795 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270795 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270795 Supplier Novus Biologicals Supplier No. NBP250057F
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

c-Fos Polyclonal antibody specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry Free-Floating

Specifications

Antigen c-Fos
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating
Formulation PBS
Gene Alias activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos
Host Species Rabbit
Immunogen This c-Fos Antibody was developed against recombinant full length human c-Fos. [UniProt# P01100]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2353
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.