Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-Fos Antibody (OTI7D6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270764H
Description
c-Fos Monoclonal specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
c-Fos | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
FOS | |
Full length recombinant protein of human c-Fos (NP_005243) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI7D6 | |
Immunohistochemistry | |
activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, c-fos, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, G0/G1 switch regulatory protein 7, G0S7, proto-oncogene c-Fos, v-fos FBJ murine osteosarcoma viral oncogene homolog | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors | |
2353.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction