Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Fos Mouse anti-Human, Mouse, Rat, Clone: 2G9C3, Novus Biologicals™
SDP

Catalog No. p-7201005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018212 0.025 mg
NB6001428 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018212 Supplier Novus Biologicals Supplier No. NB60014280.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-Fos Monoclonal specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen c-Fos
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Monoclonal
Clone 2G9C3
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.5-5 ug/ml, Immunocytochemistry/Immunofluorescence 2.5-5 ug/ml
Formulation 50mM Sodium Phosphate (pH 7.5) and 0.2% Gelatin with 0.1% Sodium Azide
Gene Accession No. P01100
Gene Alias activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, c-fos, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, G0/G1 switch regulatory protein 7, G0S7, proto-oncogene c-Fos, v-fos FBJ murine osteosarcoma viral oncogene homolog
Gene Symbols FOS
Host Species Mouse
Immunogen Synthetic peptide: KVEQLSPEEEEKRRIRRERNKMAAA, corresponding to Internal sequence amino acids 128 - 152 of Human c-Fos.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2353
Test Specificity Reacts with both cellular and viral forms, will detect both the 55 kDa and 62 kDa c-fos and v-fos proteins, but does not react with the 39 kDa jun protein.
Target Species Mouse, Human, Rat
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.