Learn More
C-Jun Mouse anti-Human, Clone: OTI2B2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800651

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5′-TGA[CG]TCA-3′. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.Specifications
C-Jun | |
Monoclonal | |
Unconjugated | |
JUN | |
AP-1, AP1, c-Jun, p39 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3725 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2B2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P05412 | |
JUN | |
Full length human recombit protein of human JUN produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.