Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-jun, p Ser63 Antibody (4A11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | c-jun |
---|---|
Clone | 4A11 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
c-jun Monoclonal antibody specifically detects c-jun in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
c-jun | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
A synthesized peptide derived from Human c-jun [p Ser63] [UniProt P05412] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4A11 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3725 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title