Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-jun, p Thr91 Antibody (SR1729), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | c-jun |
---|---|
Clone | SR1729 |
Dilution | Western Blot 1:1000-1:2000, Immunoprecipitation 1:20 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
c-jun Monoclonal antibody specifically detects c-jun in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
c-jun | |
Western Blot 1:1000-1:2000, Immunoprecipitation 1:20 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
A synthesized peptide derived from human phospho-c-Jun [Thr91] (Uniprot #: P05412) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
SR1729 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3725 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title