Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-jun, p Thr91, p Thr93 Antibody (C-J 4C4/1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002748R
Description
c-jun Monoclonal specifically detects c-jun in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
c-jun | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
JUN | |
C-Jun phosphorylated at T91 and T93 | |
0.1 mL | |
Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
3725 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
C-J 4C4/1 | |
Western Blot, ELISA | |
Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction