Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-jun Rabbit anti-Human, Clone: JJ080-9, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267858
Description
c-jun Monoclonal specifically detects c-jun in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
c-jun | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
Rabbit | |
Protein A purified | |
Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
3725 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
JJ080-9 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
JUN | |
Synthetic phospho-peptide corresponding to residues surrounding Thr91 of Human c-jun aa 60-109/ 331. (SwissProt: P05412 Human; SwissProt: P05627 Mouse; SwissProt: P17325 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction