Learn More
Invitrogen™ c-Kit (D816V Mutant) Polyclonal Antibody

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | c-Kit (D816V Mutant) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% Proclin 300; pH 7 |
| Gene Accession No. | P29466, P48729, Q8N752 |
| Gene Alias | CD117; C-Kit; KIT proto-oncogene, receptor tyrosine kinase; MASTC; PBT; SCFR |
| Host Species | Rabbit |
| Immunogen | Carrier-protein conjugated synthetic peptide surrounding mutant D816V of human c-Kit. The exact sequence is proprietary. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.