Learn More
Promega c-KIT Kinase Enzyme System, 1mg
Catalog No. PRV6579 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6579 1 mg
1 options
Catalog No. PRV6579 Supplier Promega Supplier No. V6579
Add to Cart
Add to Cart
Request a Quote
Recombinant human c-KIT (544-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag. c-KIT is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor).
  • Easily Screen and Profile c−KIT Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C