Learn More
C.L. Sturkey EXTREMUS Disposable Microtome Blades
Catalog No. p-4109279 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
Quantity:
100 Pk.
120 Pk.
170 Pk.
200 Pk.
50 Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile Quantity
22-210-034 High 100 Pk.
22-210-045 High 50 Pk.
22-210-048 Low 50 Pk.
22-210-036 Low 170 Pk.
22-210-033 High 120 Pk.
22-210-035 Low 200 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-210-034 Supplier C.L. Sturkey Supplier No. D554X
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Uniquely coated to suit the widest variety of cutting conditions
Offers a unique combination of durability and precision that saves time and money
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Profile High
Disposable Single-use
Quantity 100 Pk.