Learn More
C.L. Sturkey Silver Disposable Microtome Blades
Catalog No. 22210008
Click to view available options
Profile:
High
Low
Quantity:
170 Pk.
50 Pk.
Durable and long-lasting, ideal for general paraffin and cryostat use
Coated with a nitride and copolymer finish to provide smooth, wrinkle-free sections
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 170 Low-Profile blades per box (non-dispensing)
Specifications
Profile Low
Disposable Single-use
Quantity 170 Pk.
Coating Nitride and Copolymer
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.