Learn More
C.L. Sturkey Silver Disposable Microtome Blades
Catalog No. p-4109277 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
Quantity:
170 Pk.
50 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile Quantity
22-210-047 High 50 Pk.
22-210008 Low 170 Pk.
22-210-050 Low 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-210-047 Supplier C.L. Sturkey Supplier No. D554R50
Only null left
Add to Cart
Add to Cart
Durable and long-lasting, ideal for general paraffin and cryostat use
Coated with a nitride and copolymer finish to provide smooth, wrinkle-free sections
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 170 Low-Profile blades per box (non-dispensing)
Specifications
Profile High
Disposable Single-use
Quantity 50 Pk.
Coating Nitride and Copolymer