Learn More
Promega c-MER Kinase Enzyme System, 1mg
Catalog No. PRV3542 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3542 1 mg
1 options
Catalog No. PRV3542 Supplier Promega Supplier No. V3542
Add to Cart
Add to Cart
Request a Quote
Recombinant human c-MER (amino acids 578-872) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. c-MER is a tyrosine kinase and a member of the MER/AXL/TYRO3 receptor kinase family.
  • Easily Screen and Profile c−MER Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C