Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Met Mouse anti-Human, Brilliant Violet 510, Clone: 3D6, BD Optibuild™

Catalog No. BDB742873 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742873 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742873 Supplier BD Biosciences Supplier No. 742873
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 3D6 monoclonal antibody specifically binds to c-Met (MET), which is also known as Hepatocyte growth factor receptor (HGFR) or Scatter factor receptor (SF receptor). c-Met is a 190 kDa single-pass type I transmembrane glycoprotein that is posttranslationally cleaved into a disulfide-linked extracellularα-chain and a transmembraneβ-chain. c-Met functions as a receptor tyrosine kinase (RTK) that is normally involved in the development, regeneration, and survival of cells and tissues. c-Met is expressed on a variety of cell types including stem cells and progenitor cells, hepatocytes, keratinocytes, epithelial cells, endothelial cells, and neurons. c-Met is autophosphorylated when bound by its ligand, Hepatocyte Growth Factor (HGF). This leads to further activation of downstream signaling pathways that induce multiple responses including cellular migration, proliferation, survival, and angiogenesis. Abnormal c-Met expression or activity has been associated with tumorigenesis. The 3D6 antibody can reportedly function as an agonist by binding to and activating human but not mouse c-Met.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Met
Classification Monoclonal
Clone 3D6
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MET, cMet, HGFR, HGF receptor, AUTS9, RCCP2, SF receptor
Host Species Mouse
Immunogen Human c-Met Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.