Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ c-Met Recombinant Rabbit Monoclonal Antibody (PSH0-19)
GREENER_CHOICE

Catalog No. FEMA550789
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550789 Supplier Invitrogen™ Supplier No. MA550789
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: A549 cell lysate, HepG2 cell lysate, HT-29 cell lysate, Hela cell lysate, LO2 cell lysate, human lung adenocarcinoma tissue, Hela. Predicted band size: 156 kDa Subcellular Location: Membrane; Secreted.

MET (cMET) is a receptor-like tyrosine kinase whose dysregulation has been linked to many types of human malignancies. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Met
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PSH0-19
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MET
Gene Accession No. P08581
Gene Alias AI838057; AUTS9; bHLHe59; CG1705; CG1705-PA; CG1705-PB; cmet; c-met; c-met proto-oncogene; c-Met receptor tyrosine kinase; c-Met/HGF receptor; D249; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; HGFR; HGF-SF receptor; juvenile hormone resistance; met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; Met; protein tyrosin kinase; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; Proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; scatter factor receptor; SF receptor; soluble c met; tyrosine-protein kinase Met
Gene Symbols MET
Host Species Rabbit
Immunogen Synthetic peptide within human c-Met aa 1341-1390.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.