Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myb Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991970
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1970 100 μg
50-199-1971 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1970 Supplier Rockland Immunochemicals Supplier No. 600401B06
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with endogenous c-Myb protein and also SUMOylated overexpressed c-Myb protein in transfected Cos7 cells. A BLAST analysis was used to suggest reactivity with c-Myb from mouse based on a 100% homology with the immunizing sequence. Expect reactivity with c-Myb from rat, pig, human, horse, platypus, opossum, orangutan, spider monkey, bonobo, gorilla, and chimpanzee; with a v-myb myeloblastosis viral oncogene homolog, all isoforms (also avian version) from human; and with a protooncogene c-myb, and a v-myb myeloblastosis viral oncogene homolog (avian), both from cattle, all based on a 93% homology with the immunizing sequence. Cross-reactivity with c-Myb from other sources has not been determined.

Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5'-YAAC[GT]G-3'. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.

Specifications

Antigen c-Myb
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.51 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene MYB
Gene Accession No. P06876
Gene Alias AI550390; Cmyb; c-myb; cmyb protein; c-myb_CDS; c-myb10A_CDS; c-myb13A_CDS; c-myb14A_CDS; c-myb8B_CDS; efg; gag-myb protein; M16449; Myb; myb proto-oncogene protein; MYB proto-oncogene, transcription factor; myeloblastosis oncogene; myeloblastosis proto-oncogene product; oncogene AMV; Proto-oncogene c-Myb; RP1-32B1.1; transcription factor cmyb; Transcriptional activator Myb; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog
Gene Symbols MYB
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17863
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.