Learn More
Description
Specifications
Specifications
| Antigen | c-Myc |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 900000000000 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:200-1:400, ELISA 1:100-1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33542232), Immunoprecipitation 2ug/mg lysate, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 2 ug/ 500 ug extract, CyTOF-ready |
| Gene Accession No. | P01106 |
| Gene Alias | avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) |
| Gene Symbols | MYC |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
