Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc Mouse, HRP, Clone: 2G8D5, THE™, GenScript™

Catalog No. 502120496
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0496 Supplier Genscript Corporation Supplier No. A00863100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. GenScript THE™ c-Myc Antibody [HRP], mAb, Mouse is immunoaffinity purified from mice ascites with protein A and then conjugated to horseradish peroxidase. Mouse Anti-c-Myc [HRP] Monoclonal Antibody is suitable for detecting the expression level of c-Myc in fusion proteins.

c-Myc Monoclonal antibody specifically detects c-Myc in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen c-Myc
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2G8D5
Conjugate HRP
Formulation PBS with 1% BSA and 0.01% thimerosal; pH 7.4
Gene Alias AU016757; Avian myelocytomatosis viral (v-myc) oncogene homolog; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; mMyc; MRTL; Myc; Myc protein; Myc proto-oncogene protein; MYC proto-oncogene, bHLH transcription factor; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; myelocytomatosis viral oncogene homolog; Niard; Nird; Proto-oncogene c-Myc; RNCMYC; Transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene-like protein
Host Species Mouse
Immunogen A synthetic peptide corresponding to residues 410-419 (EQKLISEEDL) coupled to KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.