Learn More
c-Myc-tag Rabbit, Polyclonal, GenScript™
Rabbit Polyclonal Antibody
Supplier: Genscript Corporation A0017240

Description
Reconstitute the lyophilized product with deionized water (or equivalent) to the final concentration of 0.5 mg/mL. The c-Myc protein is a transcription factor, encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis, and cell cycle progression. A synthetic peptide corresponding to residues 410-419 of the human p62 c-myc protein conjugated to KLH is used as immunogen. c-Myc-tag provides a method of localizing gene products in a variety of cell types, to study the topology of proteins and protein complexes and of identifying associated proteins. This product is suitable for detecting the expression level of c-Myc fusion proteins or c-Myc by various immunoassays.
c-Myc-tag Polyclonal antibody specifically detects c-Myc-tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
c-Myc-tag | |
Polyclonal | |
PBS with 0.02% sodium azide | |
Rabbit | |
Affinity chromatography | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog | |
c-Myc epitope tag peptide EQKLISEEDL conjugated to KLH | |
40 μg | |
Primary | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.