Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-Myc tag recombinant Fab fragment, Abnova™
Recombinant Antibody
Supplier: Abnova Corporation RAB00030
Specifications
MYC | |
9E10 | |
Immunofluorescence Immunohistochemistry (Frozen sections) (5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
v-myc myelocytomatosis viral oncogene homolog (avian) | |
MYC | |
100 μg | |
Primary | |
The epitope sequence, EQKLISEEDL, is located in aa 410-419 of human c-myc protein. | |
Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
bHLHe39/c-Myc | |
Original antibody is raised against a synthetic peptide corresponding to amino acids 408-439 of human c-myc. | |
RUO | |
4609 | |
Human | |
Antibody | |
Ig F(ab) λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction