Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C-Nap1 (Cep250) Mouse anti-Human, Unconjugated, Clone: 150-230-1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT135325UG
Description
Anti-C-Nap1 (Cep250), clone 150-230-1, Cat. No. MABT1353, is a highly specific mouse monoclonal antibody that targets Centrosome-associated protein CEP250 and has been tested for use in Immunofluorescence and Western Blotting.
Specifications
| C-Nap1 (Cep250) | |
| Monoclonal | |
| Please refer to lot specific datasheet. | |
| Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| CEP250;CEP2;CNAP1 | |
| Recombinant fragment corresponding to 455 amino acids from the C-terminal region of human Centrosome-associated protein CEP250. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Clone 150-230-1 is a mouse monoclonal antibody that detects human Centrosome-associated protein CEP250 (Cep20). It targets an epitope within 455 amino acids from the C-terminal region. | |
| Stable for 1 year at 2-8°C from date of receipt. | |
| Purified |
| Immunofluorescence, Western Blot | |
| 150-230-1 | |
| Unconjugated | |
| Centrosome-associated protein CEP250;250 kDa centrosomal protein;Cep250;Centrosomal Nek2-associated protein 1;C-Nap1;Centrosomal protein 2 | |
| Mouse | |
| ∽280 kDa observed; 281.14 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
| 25 μg | |
| Epigenetics & Nuclear Function | |
| NP_001305148 | |
| Human | |
| Antibodies | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction