Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Peptide Antibody (7E10), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$203.51

Specifications

Antigen C-Peptide
Clone 70000000000
Dilution ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000, Enzyme Immunoassay, Competitive ELISA
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB172990SS
SDP
View Documents
Novus Biologicals
NB10072990SS
0.05 mg
Each of 1 for $203.51
Only null left
Add to Cart
 
Description

Description

C-Peptide Monoclonal specifically detects C-Peptide in Human samples. It is validated for ELISA, Radioimmunoassay, Enzyme Immunoassay, Competitive ELISA.
Specifications

Specifications

C-Peptide
ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000, Enzyme Immunoassay, Competitive ELISA
Monoclonal
Purified
RUO
Human
3630
Derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant C-peptide construction.
Primary
Store at 4C. Do not freeze.
70000000000
ELISA, Radioimmune Assays (RIA)
Unconjugated
Mouse
Diabetes Research
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
INS
IgG1 κ
Protein A purified
This is reacting only with human C-peptide. Binds to the human free C-peptide and C-peptide region in proinsulin molecules. Antigenic determinants identified by monoclnal antibodies are in the three-dimensional structure generated by N- and C- terminal regions of C-peptide separated by beta-turn at position 47-50 of proinsulin.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.