Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C-Peptide Antibody (CC27), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105434DL594
Description
C-Peptide Monoclonal antibody specifically detects C-Peptide in Rat samples. It is validated for ELISA, Immunoassay, ELISASpecifications
C-Peptide | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
ELISA, Immunoassay, ELISA | |
CC27 | |
ELISA, Immunoassay, Sandwich ELISA | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein. | |
0.2 mL | |
Diabetes Research, Lipid and Metabolism | |
3630 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction