Learn More
Description
Specifications
Specifications
| Antigen | C-Peptide |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoassay |
| Classification | Monoclonal |
| Clone | CC34 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoassay, Sandwich ELISA 1:100 - 1:2000 |
| Gene Alias | IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin |
| Gene Symbols | INS |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
