Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C-Reactive Protein/CRP Antibody (106), DyLight 550

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289860R
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for Sandwich ELISA Detection.Specifications
C-Reactive Protein/CRP | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
106 | |
Sandwich ELISA Detection | |
C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human C-Reactive Protein/CRP (NP_000558.2; Met1-Pro224). | |
0.1 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
1401 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction