Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C-Reactive Protein/CRP Antibody (C1), DyLight 680

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010026FR
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for Western Blot.Specifications
C-Reactive Protein/CRP | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CRP | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human CRP. | |
0.2 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
1401 | |
Human | |
Purified |
Western Blot | |
C1 | |
Western Blot | |
C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
does not react with Ca2+ depleted protein | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction